Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SelG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132535.3 | NP_572763.3 | 2 - 3 | 221 | 79 | |
GenBank mRNA | AF396454.1 | - | 2 - 3 | 225 | 79 | |
DQ285021.1 | - | 2 - 3 | 110 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |