Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3-GEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258746.2 | NP_001245675.1 | 9 - 10 | 3269 | 59 | |
NM_001347809.1 | NP_001334679.1 | 10 - 11 | 3311 | 59 | ||
NM_001347810.1 | NP_001334680.1 | 10 - 11 | 3311 | 59 | ||
NM_001347811.1 | NP_001334681.1 | 10 - 11 | 3311 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |