Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042817.3 | NP_001036282.1 | 6 - 7 | 4356 | 94 | |
NM_132955.4 | NP_573183.2 | 6 - 7 | 4482 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |