Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274508.1 | NP_001261437.1 | 3 - 4 | 838 | 102 | |
NM_001300010.1 | NP_001286939.1 | 3 - 4 | 841 | 102 | ||
NM_139688.4 | NP_647945.1 | 3 - 4 | 841 | 102 | ||
GenBank mRNA | AY071643.1 | - | 3 - 4 | 839 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |