Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blimp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274515.1 | NP_001261444.1 | 3 - 4 | 997 | 108 | |
NM_001274516.1 | NP_001261445.1 | 3 - 4 | 734 | 108 | ||
NM_001300016.1 | NP_001286945.1 | 3 - 4 | 1164 | 108 | ||
NM_139725.3 | NP_647982.1 | 3 - 4 | 1164 | 108 | ||
GenBank mRNA | AY071225.1 | - | 3 - 4 | 1164 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |