Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259733.1 | NP_001246662.1 | 2 | 1863 | 75 | |
NM_001274627.1 | NP_001261556.1 | 1 | 1276 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |