Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oseg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300053.1 | NP_001286982.1 | 2 - 3 | 1216 | 68 | |
NM_139964.3 | NP_648221.3 | 1 - 2 | 164 | 68 | ||
GenBank mRNA | BT053694.1 | - | 1 - 2 | 145 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |