Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ninaA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057308.4 | NP_476656.1 | 1 - 2 | 562 | 88 | |
GenBank mRNA | AY119494.1 | - | 1 - 2 | 522 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |