Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057354.3 | NP_476702.1 | 14 - 15 | 4386 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |