Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272239.2 | NP_001259168.1 | 3 | 2909 | 129 | |
NM_057433.4 | NP_476781.1 | 1 | 2386 | 129 | ||
GenBank mRNA | AY069393.1 | - | 1 | 2359 | 129 | |
X53417.1 | - | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |