Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vm34Ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057437.4 | NP_476785.1 | 1 | 383 | 77 | |
GenBank mRNA | BT023245.1 | - | 1 | 356 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |