Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057464.4 | NP_476812.1 | 1 - 2 | 424 | 77 | |
GenBank mRNA | AY060617.1 | - | 1 - 2 | 378 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |