Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdr65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057483.4 | NP_476831.1 | 4 - 5 | 856 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |