Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299844.1 | NP_001286773.1 | 1 - 2 | 658 | 67 | |
NM_057516.4 | NP_476864.2 | 1 - 2 | 658 | 67 | ||
NM_176254.2 | NP_788434.1 | 1 - 2 | 658 | 67 | ||
GenBank mRNA | AY118955.1 | - | 1 - 2 | 627 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |