Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299844.1 | NP_001286773.1 | 5 - 6 | 1408 | 73 | |
NM_057516.4 | NP_476864.2 | 5 - 6 | 1414 | 73 | ||
NM_176254.2 | NP_788434.1 | 5 - 6 | 1429 | 73 | ||
GenBank mRNA | AY118955.1 | - | 5 - 6 | 1398 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |