Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TfIIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273420.1 | NP_001260349.1 | 3 - 4 | 730 | 57 | |
NM_057540.4 | NP_476888.1 | 3 - 4 | 730 | 57 | ||
GenBank mRNA | BT011459.1 | - | 3 - 4 | 724 | 57 | |
BT015269.1 | - | 3 - 4 | 692 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |