Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258779.1 | NP_001245708.1 | 3 - 4 | 700 | 70 | |
NM_001258780.1 | NP_001245709.1 | 3 - 4 | 727 | 70 | ||
NM_001258781.2 | NP_001245710.1 | 3 - 4 | 829 | 70 | ||
NM_057543.4 | NP_476891.1 | 3 - 4 | 883 | 70 | ||
GenBank mRNA | AY069320.1 | - | 3 - 4 | 793 | 70 | |
L23404.1 | - | 3 - 4 | 794 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |