Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103908.2 | NP_001097378.1 | 13 - 14 | 2209 | 94 | |
NM_001299684.1 | NP_001286613.1 | 14 - 15 | 2254 | 94 | ||
NM_057699.5 | NP_477047.1 | 13 - 14 | 2117 | 94 | ||
GenBank mRNA | AY051678.1 | - | 12 - 13 | 2093 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |