Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057732.4 | NP_477080.1 | 3 - 4 | 842 | 80 | |
GenBank mRNA | AY061049.1 | - | 3 - 4 | 799 | 80 | |
X99511.1 | - | 818 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |