Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi3K59F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057785.5 | NP_477133.1 | 3 - 4 | 960 | 73 | |
GenBank mRNA | AY058362.1 | - | 3 - 4 | 937 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |