Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi3K59F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057785.5 | NP_477133.1 | 6 - 7 | 2487 | 67 | |
GenBank mRNA | AY058362.1 | - | 6 - 7 | 2464 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |