Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298940.1 | NP_001285869.1 | 1 | 1758 | 96 | |
NM_057824.5 | NP_477172.1 | 1 | 1758 | 96 | ||
GenBank mRNA | AY060261.1 | - | 1 | 1766 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |