Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273947.1 | NP_001260876.1 | 1 - 2 | 255 | 70 | |
NM_057859.4 | NP_477207.1 | 1 - 2 | 554 | 70 | ||
GenBank mRNA | BT003445.1 | - | 1 - 2 | 256 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |