Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272885.1 | NP_001259814.1 | 1 - 2 | 238 | 71 | |
NM_057860.4 | NP_477208.1 | 1 - 2 | 238 | 71 | ||
GenBank mRNA | AY058537.1 | - | 1 - 2 | 212 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |