Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103770.2 | NP_001097240.1 | 3 - 4 | 1332 | 67 | |
NM_001259295.2 | NP_001246224.1 | 3 - 4 | 1332 | 67 | ||
NM_001273896.1 | NP_001260825.1 | 3 - 4 | 1332 | 67 | ||
NM_057867.4 | NP_477215.1 | 3 - 4 | 1332 | 67 | ||
GenBank mRNA | AY075385.1 | - | 3 - 4 | 1299 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |