Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057909.5 | NP_477257.2 | 1 - 2 | 176 | 96 | |
GenBank mRNA | AY060686.1 | - | 1 - 2 | 152 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |