Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057963.5 | NP_477311.1 | 2 - 3 | 1560 | 62 | |
NM_164485.2 | NP_722810.1 | 2 - 3 | 1014 | 62 | ||
NM_164486.2 | NP_722811.1 | 2 - 3 | 2067 | 62 | ||
NM_164488.2 | NP_722813.1 | 2 - 3 | 1053 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |