Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057966.5 | NP_477314.1 | 2 - 3 | 149 | 71 | |
GenBank mRNA | AY071712.1 | - | 2 - 3 | 149 | 71 | |
KX532110.1 | - | 1 - 2 | 108 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |