Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258605.1 | NP_001245534.1 | 5 - 6 | 692 | 53 | |
NM_001297972.1 | NP_001284901.1 | 5 - 6 | 827 | 53 | ||
NM_080053.4 | NP_524792.1 | 5 - 6 | 827 | 53 | ||
GenBank mRNA | AB042625.2 | - | 5 - 6 | 745 | 53 | |
AY070980.1 | - | 5 - 6 | 825 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |