Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103847.2 | NP_001097317.1 | 3 - 4 | 800 | 68 | |
NM_001274051.1 | NP_001260980.1 | 2 - 3 | 469 | 68 | ||
NM_080074.4 | NP_524813.2 | 2 - 3 | 921 | 68 | ||
NM_137146.3 | NP_610990.2 | 2 - 3 | 921 | 68 | ||
NM_166070.3 | NP_725419.2 | 2 - 3 | 921 | 68 | ||
GenBank mRNA | BT150395.1 | - | 2 - 3 | 919 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |