Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080091.4 | NP_524830.2 | 1 - 2 | 198 | 67 | |
GenBank mRNA | AY069144.1 | - | 1 - 2 | 179 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |