Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S6KL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258813.1 | NP_001245742.1 | 2 - 3 | 254 | 69 | |
NM_001258814.2 | NP_001245743.1 | 2 - 3 | 726 | 69 | ||
NM_080122.4 | NP_524861.2 | 2 - 3 | 732 | 69 | ||
GenBank mRNA | AY069491.1 | - | 2 - 3 | 626 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |