Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ebo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297809.1 | NP_001284738.1 | 2 - 3 | 704 | 62 | |
NM_001297810.1 | NP_001284739.1 | 2 - 3 | 704 | 62 | ||
NM_080140.3 | NP_524879.2 | 2 - 3 | 704 | 62 | ||
GenBank mRNA | BT132921.1 | - | 2 - 3 | 672 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |