Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SrpRalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080148.3 | NP_524887.2 | 2 - 3 | 870 | 61 | |
GenBank mRNA | BT044198.1 | - | 2 - 3 | 855 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |