Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spn38F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299183.1 | NP_001286112.1 | 2 - 3 | 861 | 69 | |
NM_080217.4 | NP_524956.2 | 2 - 3 | 861 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |