Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273452.1 | NP_001260381.1 | 5 - 6 | 1833 | 81 | |
NM_080254.3 | NP_524993.3 | 5 - 6 | 1833 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |