Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297906.1 | NP_001284835.1 | 4 - 5 | 1242 | 78 | |
NM_080319.5 | NP_525058.2 | 4 - 5 | 1242 | 78 | ||
GenBank mRNA | BT057980.1 | - | 4 - 5 | 1240 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |