Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297925.1 | NP_001284854.1 | 9 - 10 | 2955 | 73 | |
NM_080327.3 | NP_525066.1 | 9 - 10 | 2919 | 73 | ||
GenBank mRNA | M77166.1 | - | 9 - 10 | 2860 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |