Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259641.1 | NP_001246570.1 | 3 - 4 | 2170 | 77 | |
NM_001259642.1 | NP_001246571.1 | 3 - 4 | 2170 | 77 | ||
NM_001259643.1 | NP_001246572.1 | 3 - 4 | 2170 | 77 | ||
NM_001259644.1 | NP_001246573.1 | 3 - 4 | 2170 | 77 | ||
NM_001259645.1 | NP_001246574.1 | 3 - 4 | 2170 | 77 | ||
NM_001259646.1 | NP_001246575.1 | 3 - 4 | 2170 | 77 | ||
NM_001259647.1 | NP_001246576.1 | 2 - 3 | 1578 | 77 | ||
NM_001259648.1 | NP_001246577.1 | 3 - 4 | 2170 | 77 | ||
NM_001259649.1 | NP_001246578.1 | 3 - 4 | 2170 | 77 | ||
NM_001299992.1 | NP_001286921.1 | 3 - 4 | 2170 | 77 | ||
NM_080516.3 | NP_536777.3 | 3 - 4 | 2170 | 77 | ||
GenBank mRNA | BT133165.1 | - | 3 - 4 | 2166 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |