Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201735.2 | NP_001188664.1 | 2 - 3 | 678 | 61 | |
NM_133018.4 | NP_573246.1 | 2 - 3 | 738 | 61 | ||
GenBank mRNA | BT023820.1 | - | 2 - 3 | 700 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |