Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mur18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133136.3 | NP_573364.1 | 1 - 2 | 157 | 72 | |
GenBank mRNA | BT004882.1 | - | 1 - 2 | 140 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |