Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169348.2 | NP_001162819.1 | 3 - 4 | 281 | 117 | |
NM_175926.3 | NP_787122.1 | 2 - 3 | 239 | 117 | ||
NM_175927.3 | NP_787123.1 | 3 - 4 | 314 | 117 | ||
NM_175928.3 | NP_787124.1 | 3 - 4 | 429 | 117 | ||
GenBank mRNA | BT031123.1 | - | 2 - 3 | 234 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |