Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058057.6 | NP_477405.1 | 1 - 2 | 267 | 65 | |
GenBank mRNA | AF071103.1 | - | 1 - 2 | 270 | 65 | |
BT001772.1 | - | 1 - 2 | 257 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |