Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274452.1 | NP_001261381.1 | 2 - 3 | 636 | 90 | |
NM_133165.2 | NP_573393.1 | 2 - 3 | 793 | 90 | ||
GenBank mRNA | AY060992.1 | - | 2 - 3 | 614 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |