Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyc32E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103678.2 | NP_001097148.1 | 13 - 14 | 1889 | 72 | |
NM_001298905.1 | NP_001285834.1 | 12 - 13 | 1781 | 72 | ||
NM_176013.2 | NP_788027.1 | 12 - 13 | 1805 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |