Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144216.3 | NP_001137688.2 | 2 - 3 | 1347 | 63 | |
NM_001144217.2 | NP_001137689.2 | 2 - 3 | 1347 | 63 | ||
NM_001259462.2 | NP_001246391.1 | 2 - 3 | 1189 | 63 | ||
NM_001259464.2 | NP_001246393.1 | 2 - 3 | 1347 | 63 | ||
NM_001259466.2 | NP_001246395.1 | 2 - 3 | 1189 | 63 | ||
NM_001274114.1 | NP_001261043.1 | 2 - 3 | 1347 | 63 | ||
NM_001274115.1 | NP_001261044.1 | 3 - 4 | 1229 | 63 | ||
NM_001274117.1 | NP_001261046.1 | 2 - 3 | 1347 | 63 | ||
NM_001274118.1 | NP_001261047.1 | 2 - 3 | 1347 | 63 | ||
NM_001274119.1 | NP_001261048.1 | 2 - 3 | 1347 | 63 | ||
NM_176210.2 | NP_788390.1 | 2 - 3 | 1347 | 63 | ||
NM_176211.2 | NP_788391.1 | 2 - 3 | 1347 | 63 | ||
NM_176212.2 | NP_788392.1 | 2 - 3 | 1347 | 63 | ||
GenBank mRNA | AF001536.1 | - | 1 - 2 | 348 | 63 | |
AF001625.1 | - | 2 - 3 | 1329 | 63 | ||
JN812971.1 | - | 1 - 2 | 188 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |