Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp65Ab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176291.3 | NP_788469.1 | 1 | 349 | 65 | |
GenBank mRNA | AY071366.1 | - | 1 | 346 | 65 |
Gene Symbol: Lcp65Ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080075.3 | NP_524814.2 | 1 | 346 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |