Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176699.3 | NP_788872.1 | 1 - 2 | 428 | 71 | |
GenBank mRNA | BT099510.1 | - | 1 - 2 | 423 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |