Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298345.1 | NP_001285274.1 | 2 - 3 | 3195 | 67 | |
NM_001370008.1 | NP_001356946.1 | 3 - 4 | 3527 | 67 | ||
NM_176734.2 | NP_788907.1 | 3 - 4 | 3527 | 67 | ||
NM_176735.3 | NP_788908.1 | 2 - 3 | 3335 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |