Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014613.3 | NP_001014613.2 | 4 - 5 | 2188 | 77 | |
NM_001031999.2 | NP_001027170.1 | 3 - 4 | 1719 | 77 | ||
NM_057228.5 | NP_476576.1 | 3 - 4 | 1719 | 77 | ||
NM_057229.5 | NP_476577.3 | 3 - 4 | 1719 | 77 | ||
NM_057230.6 | NP_476578.3 | 3 - 4 | 1719 | 77 | ||
GenBank mRNA | AF032865.1 | - | 3 - 4 | 813 | 77 | |
AF035825.1 | - | 3 - 4 | 1695 | 77 | ||
AF036584.1 | - | 3 - 4 | 900 | 77 | ||
BT024210.1 | - | 3 - 4 | 1538 | 77 | ||
BT149862.1 | - | 4 - 5 | 1284 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |